siRNA Details
Gene Symbol: Vps8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285893.1 | NP_001272822.1 | 23 | 2399 | ||
NM_001285894.1 | NP_001272823.1 | 24 | 2352 | |||
GenBank mRNA | AK029007.1 | - | 7 | 616 | ||
AK122375.1 | - | 24 | 2301 | |||
AK141678.1 | - | 23 | 2322 | |||
BC023914.1 | - | 9 | 790 | 5323126 | ||
BC032214.1 | - | 3 | 368 | 5323693 | ||
BC055323.1 | - | 23 | 2316 | 5706288 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
