siRNA Details
Gene Symbol: Serinc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025371.2 | NP_001020542.1 | 5 | 769 | ||
GenBank mRNA | AK044479.1 | - | 5 | 797 | ||
BC026459.1 | - | 5 | 698 | |||
BC083132.1 | - | 5 | 769 | 6466398 |

More Information
Other Species Targeted

