siRNA Details
Gene Symbol: Cep164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081373.2 | NP_001074842.2 | 8 | 1145 | ||
GenBank mRNA | AK032890.1 | - | 3 | 306 | ||
AK220365.1 | - | 4 | 491 | |||
BC027092.1 | - | 1 | 363 | |||
BC076567.1 | - | 6 | 886 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
