siRNA Details
Gene Symbol: Vps8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285893.1 | NP_001272822.1 | 24, 25 | 2465 | ||
NM_001285894.1 | NP_001272823.1 | 25, 26 | 2418 | |||
GenBank mRNA | AK029007.1 | - | 8, 9 | 682 | ||
AK122375.1 | - | 25, 26 | 2367 | |||
AK141678.1 | - | 24, 25 | 2388 | |||
BC023914.1 | - | 10, 11 | 856 | 5323126 | ||
BC032214.1 | - | 4, 5 | 434 | 5323693 | ||
BC055323.1 | - | 24, 25 | 2382 | 5706288 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
