siRNA Details
Gene Symbol: Serf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BP768998.1 | - | 1 | 360 |
Gene Symbol: Serinc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025371.2 | NP_001020542.1 | 9 | 1323 | ||
GenBank mRNA | AK044479.1 | - | 9 | 1351 | ||
BC026459.1 | - | 7 | 1029 | |||
BC083132.1 | - | 9 | 1323 | 6466398 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
