siRNA Details
Gene Symbol: Vps8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285893.1 | NP_001272822.1 | 26, 27 | 2596 | ||
NM_001285894.1 | NP_001272823.1 | 27, 28 | 2549 | |||
GenBank mRNA | AK029007.1 | - | 10, 11 | 813 | ||
AK031205.1 | - | 3, 4 | 637 | |||
AK122375.1 | - | 27, 28 | 2498 | |||
AK141678.1 | - | 26, 27 | 2519 | |||
BC023914.1 | - | 12, 13 | 987 | 5323126 | ||
BC032214.1 | - | 6, 7 | 565 | 5323693 | ||
BC055323.1 | - | 26, 27 | 2513 | 5706288 |

More Information
Other Species Targeted

Gene Ontology Categories:
