siRNA Details
Gene Symbol: Serinc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025371.2 | NP_001020542.1 | 12 | 2196 | ||
GenBank mRNA | BC026459.1 | - | 10 | 1902 | ||
BC083132.1 | - | 12 | 2196 | 6466398 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
