siRNA Details
Gene Symbol: Larp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040399.1 | NP_001035489.1 | 9 | 999 | ||
GenBank mRNA | AK076732.1 | - | 9 | 967 | ||
AK160155.1 | - | 9 | 999 | |||
AK163348.1 | - | 8 | 965 | |||
AK212523.1 | - | 2 | 131 | |||
BC025528.1 | - | 7 | 922 | 5324328 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
