siRNA Details
Gene Symbol: Kdm5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145997.2 | NP_666109.2 | 1, 2 | 688 | ||
GenBank mRNA | AK041568.1 | - | 1, 2 | 688 | ||
AK046187.1 | - | 1, 2 | 638 | |||
AK133589.1 | - | 1, 2 | 357 | |||
AK139619.1 | - | 1, 2 | 655 | |||
AK144877.1 | - | 1, 2 | 659 | |||
AK154025.1 | - | 1, 2 | 344 | |||
AK159079.1 | - | 1, 2 | 665 | |||
AK166055.1 | - | 1, 2 | 669 | |||
BC010717.1 | - | 1, 2 | 329 | |||
BC011506.3 | - | 1, 2 | 622 | |||
BC080691.1 | - | 1, 2 | 374 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
