siRNA Details
Gene Symbol: Cep290 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146009.2 | NP_666121.2 | 37 | 5066 | ||
GenBank mRNA | AK029960.1 | - | 2 | 73 | ||
AK172940.1 | - | 10 | 1722 | |||
BC004690.1 | - | 3 | 352 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
