siRNA Details
Gene Symbol: Chd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146019.4 | NP_666131.3 | 35 | 5430 | ||
GenBank mRNA | BC030435.1 | - | 2 | 313 | ||
BM950040.1 | - | 5 | 547 | |||
CB195281.1 | - | 5 | 611 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
