siRNA Details
Gene Symbol: Ina | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146100.4 | NP_666212.3 | 2 | 1153 | ||
GenBank mRNA | BC018383.1 | - | 2 | 1153 | 4502421 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
