siRNA Details
Gene Symbol: Aph1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146104.3 | NP_666216.1 | 2 | 507 | ||
NM_146134.2 | NP_666246.1 | 2 | 507 | |||
GenBank mRNA | AK050828.1 | - | 1 | 500 | ||
AK148400.1 | - | 2 | 460 | |||
AK152334.1 | - | 2 | 450 | |||
AX775953.1 | - | 2 | 375 | |||
BC012406.1 | - | 2 | 432 | 3483214 | ||
BC024111.1 | - | 2 | 373 | 5098973 | ||
BC057865.1 | - | 2 | 450 | 5320175 |
