siRNA Details
Gene Symbol: Chd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146019.4 | NP_666131.3 | 37, 38 | 5791 | ||
GenBank mRNA | BC030435.1 | - | 4, 5 | 674 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008767924, AY724529, AY903245, AY903250, AY903251, and others.
Gene Ontology Categories:
