siRNA Details
Gene Symbol: Ccar2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146055.3 | NP_666167.3 | 21 | 3156 | ||
GenBank mRNA | AK028194.1 | - | 21 | 3076 | ||
AK031472.1 | - | 17 | 2696 | |||
BC010499.1 | - | 3 | 2042 | |||
BC021475.1 | - | 21 | 3186 | 5343456 | ||
BC025471.1 | - | 8 | 1378 | |||
BC038346.1 | - | 21 | 3108 | 3985250 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
