siRNA Details
Gene Symbol: Krt79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146063.1 | NP_666175.1 | 1 | 454 | ||
GenBank mRNA | BC019155.1 | - | 1 | 393 | 5065687 | |
BC031593.1 | - | 1 | 454 | 4935395 |

More Information
Other Species Targeted

Gene Ontology Categories:
