siRNA Details
Gene Symbol: Ermp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081213.1 | NP_001074682.1 | 12, 13 | 2325 | ||
GenBank mRNA | AK050284.1 | - | 12, 13 | 2322 | ||
AK081674.1 | - | 11, 12 | 1842 | |||
AK129447.1 | - | 12, 13 | 2253 | |||
AK137206.1 | - | 12, 13 | 2281 | |||
BC031519.1 | - | 4, 5 | 601 | |||
BC151181.1 | - | 12, 13 | 2284 | 9088070 | ||
BC151182.1 | - | 12, 13 | 2284 | 9088071 | ||
BC157914.1 | - | 12, 13 | 2284 | 9088152 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_184050, XM_006231241, AY364634.
Gene Ontology Categories:
