siRNA Details
Gene Symbol: Tmem185b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146103.2 | NP_666215.1 | 1 | 841 | ||
GenBank mRNA | AK010846.1 | - | 1 | 569 | ||
AK031130.1 | - | 1 | 837 | |||
BC020408.1 | - | 1 | 829 | |||
BC024712.1 | - | 1 | 828 | 2646072 |
