siRNA Details
Gene Symbol: Kdm5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145997.2 | NP_666109.2 | 4 | 905 | ||
GenBank mRNA | AK041568.1 | - | 4 | 905 | ||
AK046187.1 | - | 4 | 855 | |||
AK133589.1 | - | 4 | 574 | |||
AK144877.1 | - | 4 | 876 | |||
AK154025.1 | - | 4 | 561 | |||
AK159079.1 | - | 4 | 882 | |||
AK166055.1 | - | 4 | 886 | |||
BC010717.1 | - | 4 | 546 | |||
BC011506.3 | - | 4 | 839 | |||
BC080691.1 | - | 4 | 591 |

More Information
Other Species Targeted

Gene Ontology Categories:
