siRNA Details
Gene Symbol: Cep290 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146009.2 | NP_666121.2 | 52 | 7175 | ||
GenBank mRNA | AK029960.1 | - | 17 | 2182 | ||
AK172305.1 | - | 160 | ||||
AK172940.1 | - | 25 | 3831 | |||
BC004690.1 | - | 15 | 1978 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
