siRNA Details
Gene Symbol: Ccar2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146055.3 | NP_666167.3 | 21 | 3326 | ||
GenBank mRNA | AK028194.1 | - | 21 | 3246 | ||
AK031472.1 | - | 17 | 2866 | |||
BC010499.1 | - | 3 | 2212 | |||
BC021475.1 | - | 21 | 3356 | 5343456 | ||
BC025471.1 | - | 8 | 1548 | |||
BC038346.1 | - | 21 | 3278 | 3985250 |

More Information
Other Species Targeted

Gene Ontology Categories:
