siRNA Details
Gene Symbol: Gigyf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110212.2 | NP_001103682.1 | 29 | 5056 | ||
NM_146112.4 | NP_666224.3 | 29 | 5074 | |||
GenBank mRNA | AK044009.1 | - | 1 | 272 | ||
AK122337.1 | - | 20 | 4145 | |||
BC025466.1 | - | 4 | 1504 | |||
BC027137.1 | - | 9 | 2418 | |||
BC030845.1 | - | 4 | 1516 | |||
BC076610.1 | - | 29 | 5019 | |||
BC082811.1 | - | 11 | 2475 | |||
BC089036.1 | - | 29 | 5030 | 30695300 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
