siRNA Details
Gene Symbol: Hnrnpa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053263.1 | NP_444493.1 | 11 | 1186 | ||
NM_146130.3 | NP_666242.2 | 11 | 1252 | |||
GenBank mRNA | AB201711.1 | - | 11 | 1232 | ||
AF463524.1 | - | 11 | 1164 | |||
AK133860.1 | - | 11 | 1251 | |||
AK169663.1 | - | 11 | 1234 | |||
BC023828.1 | - | 11 | 1205 | 5343619 | ||
BC023908.1 | - | 11 | 1221 | 5322033 | ||
BC024454.1 | - | 8 | 769 | |||
BC062198.1 | - | 11 | 1060 | 2655879 | ||
BC085474.1 | - | 10 | 1127 | 30538435 | ||
BC092083.1 | - | 8 | 847 | |||
BC157919.1 | - | 11 | 1155 | 9007347 | ||
BC158038.1 | - | 11 | 1155 | 9007341 |
Gene Symbol: Gm6793 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC157904.1 | - | 1 | 1137 | 9088095 | |
BC158074.1 | - | 1 | 1137 | 9007371 | ||
BC172011.1 | - | 1 | 1137 | 9054297 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
