siRNA Details
Gene Symbol: Dlgap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042487.1 | NP_001035952.1 | 2 | 1169 | ||
NM_001042488.2 | NP_001035953.1 | 2 | 468 | |||
NM_001277186.1 | NP_001264115.1 | 8 | 2413 | |||
NM_001277187.1 | NP_001264116.1 | 7 | 2362 | |||
NM_146128.6 | NP_666240.4 | 8 | 2413 | |||
GenBank mRNA | AK089484.1 | - | 2 | 389 | ||
AK122408.1 | - | 8 | 2413 | |||
AK150554.1 | - | 2 | 389 | |||
AK156212.1 | - | 2 | 389 | |||
AY243849.2 | - | 6 | 1993 | |||
BC024558.1 | - | 2 | 573 | |||
BC058948.1 | - | 2 | 492 | |||
BC085475.1 | - | 2 | 956 | |||
BC106094.1 | - | 2 | 551 | |||
BC141110.1 | - | 6 | 2050 | 9055722 | ||
BC145394.1 | - | 6 | 2050 | 9053952 | ||
BC145395.1 | - | 5 | 1999 | 9053953 |

More Information
Other Species Targeted

Gene Ontology Categories:
