siRNA Details
Gene Symbol: Hnrnpa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053263.1 | NP_444493.1 | 11 | 1207 | ||
NM_146130.3 | NP_666242.2 | 11 | 1273 | |||
GenBank mRNA | AB201711.1 | - | 11 | 1253 | ||
AK133860.1 | - | 11 | 1272 | |||
AK169663.1 | - | 11 | 1255 | |||
BC023828.1 | - | 11 | 1226 | 5343619 | ||
BC023908.1 | - | 11 | 1242 | 5322033 | ||
BC024454.1 | - | 8 | 790 | |||
BC062198.1 | - | 11 | 1081 | 2655879 | ||
BC085474.1 | - | 10 | 1148 | 30538435 | ||
BC092083.1 | - | 8 | 868 | |||
BC157919.1 | - | 11 | 1176 | 9007347 | ||
BC158038.1 | - | 11 | 1176 | 9007341 |
Gene Symbol: Gm6793 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC157904.1 | - | 1 | 1158 | 9088095 | |
BC158074.1 | - | 1 | 1158 | 9007371 | ||
BC172011.1 | - | 1 | 1158 | 9054297 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001111294, XM_008761913, XM_008761911, Y16641, CF110891, and others.
Gene Ontology Categories:
