siRNA Details
Gene Symbol: Myo1h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164573.1 | NP_001158045.1 | 25 | 2543 | ||
GenBank mRNA | AF426467.1 | - | 3 | 224 | ||
AK014505.1 | - | 4 | 473 | |||
BC144870.1 | - | 4 | 424 | 9053418 | ||
BC144872.1 | - | 4 | 448 |
