siRNA Details
Gene Symbol: Tcaf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146174.1 | NP_666286.1 | 2 | 547 | ||
GenBank mRNA | AK131169.1 | - | 2 | 760 | ||
BC008555.1 | - | 2 | 564 | 3592166 | ||
BC011487.1 | - | 2 | 547 | 3499593 |
Gene Symbol: Tcaf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203396.1 | NP_981933.1 | 1 | 467 | ||
GenBank mRNA | AY528666.1 | - | 1 | 467 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
