siRNA Details
Gene Symbol: Wdr62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146186.3 | NP_666298.3 | 24 | 3001 | ||
GenBank mRNA | AK154592.1 | - | 24 | 2972 | ||
BC026444.1 | - | 5 | 568 | |||
BC054747.1 | - | 24 | 2976 | 6405269 | ||
BC057041.1 | - | 14 | 1511 | 6817055 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
