siRNA Details
Gene Symbol: Zfp825 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146231.1 | NP_666343.1 | 4 | 1152 | ||
GenBank mRNA | BC012278.1 | - | 4 | 1152 | 3985027 | |
BC066158.1 | - | 6 | 1201 | |||
BC094048.1 | - | 4 | 1206 | 5251253 | ||
BC119319.1 | - | 4 | 1097 | 8734068 |
