siRNA Details
Gene Symbol: Dlgap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042487.1 | NP_001035952.1 | 7 | 2173 | ||
NM_001042488.2 | NP_001035953.1 | 7 | 1472 | |||
NM_001277186.1 | NP_001264115.1 | 13 | 3408 | |||
NM_001277187.1 | NP_001264116.1 | 12 | 3366 | |||
NM_146128.6 | NP_666240.4 | 13 | 3417 | |||
GenBank mRNA | AK122408.1 | - | 11 | 6075 | ||
AK150554.1 | - | 7 | 1393 | |||
AK156212.1 | - | 5 | 4051 | |||
AK179758.1 | - | 1 | 66 | |||
BC024558.1 | - | 7 | 1577 | |||
BC058948.1 | - | 7 | 1496 | |||
BC085475.1 | - | 7 | 1960 | |||
BC106094.1 | - | 7 | 1555 | |||
BC141110.1 | - | 11 | 3045 | 9055722 | ||
BC145394.1 | - | 11 | 3045 | 9053952 | ||
BC145395.1 | - | 10 | 3003 | 9053953 |
