siRNA Details
Gene Symbol: Fam151a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146149.1 | NP_666261.1 | 8 | 1878 | ||
GenBank mRNA | AK008396.1 | - | 1 | 431 | ||
AK008553.1 | - | 1 | 431 | |||
AY168619.1 | - | 8 | 1878 | |||
BC024426.1 | - | 8 | 1878 | 4971419 | ||
BC024430.1 | - | 8 | 1878 | 4972368 | ||
BC026682.1 | - | 8 | 1887 | 5065146 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
