siRNA Details
Gene Symbol: Zfp764 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167832.1 | NP_001161304.1 | 3 | 2419 | ||
NM_146203.4 | NP_666315.3 | 3 | 2456 | |||
GenBank mRNA | AK042873.1 | - | 3 | 2457 | ||
AK053795.1 | - | 3 | 2381 | |||
BC026397.1 | - | 3 | 2341 | 4207307 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
