siRNA Details
Gene Symbol: Aars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146217.4 | NP_666329.2 | 3709 | |||
GenBank mRNA | AK208535.1 | - | 100 | |||
BC013261.1 | - | 2934 | ||||
BC026611.1 | - | 1738 | ||||
BC058620.1 | - | 1883 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
