siRNA Details
Gene Symbol: Zfp119b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146249.4 | NP_666361.1 | 3 | 252 | ||
GenBank mRNA | BC031441.1 | - | 3 | 252 | 3985007 | |
BC137918.1 | - | 3 | 235 | 8860937 | ||
BC137919.1 | - | 3 | 235 | 8860938 |
