siRNA Details
Gene Symbol: Olfr1274-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146263.2 | NP_666375.2 | 1 | 914 | ||
GenBank mRNA | BC145937.1 | - | 914 | 40131240 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
