siRNA Details
Gene Symbol: Olfr846 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146282.2 | NP_666394.1 | 1 | 275 | ||
GenBank mRNA | BC134375.1 | - | 1 | 276 | 40135046 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
