siRNA Details
Gene Symbol: Olfr113 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146289.1 | NP_666401.1 | 1 | 474 | ||
GenBank mRNA | BC132178.1 | - | 1 | 501 | 40130455 | |
BC132180.1 | - | 1 | 501 | 40130457 |
Gene Symbol: Olfr116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146632.1 | NP_666843.1 | 1 | 474 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
