siRNA Details
Gene Symbol: Olfr1143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146293.2 | NP_666405.1 | 1 | 86 | ||
GenBank mRNA | CB173239.1 | - | 1 | 172 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146293.2 | NP_666405.1 | 1 | 86 | ||
GenBank mRNA | CB173239.1 | - | 1 | 172 |