siRNA Details
Gene Symbol: Olfr1167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146294.2 | NP_666406.2 | 1 | 230 | ||
GenBank mRNA | BC119292.1 | - | 1 | 230 | 8734041 | |
BC137817.1 | - | 1 | 230 | 8860836 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
