siRNA Details
Gene Symbol: Olfr601 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146314.2 | NP_666426.2 | 1 | 206 | ||
GenBank mRNA | BC131956.1 | - | 1 | 206 | 40130233 | |
BC131958.1 | - | 1 | 206 | 40130235 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
