siRNA Details
Gene Symbol: Olfr1232 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146323.1 | NP_666435.1 | 1 | 291 |
Gene Symbol: Olfr1229 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011761.1 | NP_001011761.1 | 1 | 291 | ||
GenBank mRNA | BC147797.1 | - | 1 | 317 | 9087514 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
