siRNA Details
Gene Symbol: Olfr809 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146324.1 | NP_666436.1 | 1 | 311 | ||
GenBank mRNA | BC120625.1 | - | 1 | 355 | 40129548 | |
BC137816.1 | - | 1 | 355 | 8860835 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
