siRNA Details
Gene Symbol: Olfr954 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146331.1 | NP_666443.1 | 1 | 718 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001000966.
Gene Ontology Categories:
