siRNA Details
Gene Symbol: Zfp119b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146249.4 | NP_666361.1 | 4 | 1143 | ||
GenBank mRNA | BC031441.1 | - | 4 | 1143 | 3985007 | |
BC094043.1 | - | 1332 | 5067417 | |||
BC137918.1 | - | 4 | 1126 | 8860937 | ||
BC137919.1 | - | 4 | 1126 | 8860938 |
