siRNA Details
Gene Symbol: Olfr1511 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146271.2 | NP_666383.2 | 1 | 388 | ||
GenBank mRNA | BC112968.1 | - | 1 | 386 |
Gene Symbol: Olfr1510 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146431.2 | NP_666642.2 | 1 | 445 | ||
GenBank mRNA | BC112967.1 | - | 388 | 40043789 | ||
BC114920.2 | - | 388 | 40040851 | |||
BC119377.1 | - | 1 | 445 | 8734126 | ||
BC148301.2 | - | 388 | 40043795 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
