siRNA Details
Gene Symbol: Olfr1311 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146274.1 | NP_666386.1 | 1 | 713 | ||
GenBank mRNA | BC127967.1 | - | 1 | 717 | 40135022 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
