siRNA Details
Gene Symbol: Olfr960 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146279.1 | NP_666391.1 | 1 | 527 | ||
GenBank mRNA | BC132468.1 | - | 1 | 547 | 40130745 | |
BC132470.1 | - | 1 | 547 | 40130747 |
Gene Symbol: Olfr961 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146504.1 | NP_666715.1 | 1 | 521 | ||
GenBank mRNA | BC127985.1 | - | 1 | 525 | 40135556 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
