siRNA Details
Gene Symbol: PKN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317926.1 | NP_001304855.1 | 20 | 2718 | ||
NM_013355.4 | NP_037487.2 | 20 | 2718 | |||
XM_005251946.3 | XP_005252003.1 | 20 | 3273 | |||
XM_006717080.2 | XP_006717143.1 | 20 | 2787 | |||
XM_017014649.1 | XP_016870138.1 | 20 | 2789 | |||
XM_017014650.1 | XP_016870139.1 | 20 | 2927 | |||
GenBank mRNA | AB019692.1 | - | 20 | 2306 | ||
BC062558.1 | - | 20 | 2321 | 6649806 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
