siRNA Details
Gene Symbol: Olfr1167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146294.2 | NP_666406.2 | 1 | 985 | ||
GenBank mRNA | BC119292.1 | - | 1 | 985 | 8734041 | |
BC137817.1 | - | 1 | 985 | 8860836 |
Gene Symbol: Olfr1166 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146650.2 | NP_666861.2 | 1 | 929 | ||
GenBank mRNA | BC051250.1 | - | 1 | 929 | 6306097 |
Gene Symbol: Olfr1170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146532.1 | NP_666743.1 | 1 | 919 | ||
GenBank mRNA | BC127994.1 | - | 1 | 923 | 40135743 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
