siRNA Details
Gene Symbol: Olfr1113 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207565.1 | NP_997448.1 | 1 | 750 |
Gene Symbol: Olfr1115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146297.2 | NP_666409.2 | 1 | 849 | ||
GenBank mRNA | BC141024.1 | - | 1 | 849 | 9055636 |

More Information
Other Species Targeted

Gene Ontology Categories:
